Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGTGCCAAGGCCTCTTCCGCTGT[A/G]TAGCGGTTCTGGGGTTGCACCACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 172470 MIM: 607940 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHKG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PHKG1 - phosphorylase kinase catalytic subunit gamma 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258459.1 | 999 | Silent Mutation | NP_001245388.1 | |||
NM_001258460.1 | 999 | Silent Mutation | NP_001245389.1 | |||
NM_006213.4 | 999 | Silent Mutation | NP_006204.1 | |||
XM_005271772.4 | 999 | Silent Mutation | XP_005271829.1 | |||
XM_017012324.1 | 999 | Silent Mutation | XP_016867813.1 | |||
XM_017012325.1 | 999 | Nonsense Mutation | XP_016867814.1 | |||
XM_017012326.1 | 999 | Silent Mutation | XP_016867815.1 | |||
XM_017012327.1 | 999 | Silent Mutation | XP_016867816.1 |
SUMF2 - sulfatase modifying factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |