Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGGCGGGGGAGTCTTCACATTTG[C/T]CATCCCCCTCATCCTGGGCAGCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611309 MIM: 610559 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C10orf10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C10orf10 - chromosome 10 open reading frame 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASSF4 - Ras association domain family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032023.3 | 247 | Intron | NP_114412.2 | |||
XM_005271833.4 | 247 | Intron | XP_005271890.1 | |||
XM_005271835.4 | 247 | Intron | XP_005271892.1 | |||
XM_006718013.3 | 247 | Intron | XP_006718076.1 | |||
XM_006718014.3 | 247 | UTR 5 | XP_006718077.1 | |||
XM_011540255.2 | 247 | Intron | XP_011538557.1 | |||
XM_017016754.1 | 247 | UTR 5 | XP_016872243.1 | |||
XM_017016755.1 | 247 | UTR 5 | XP_016872244.1 | |||
XM_017016756.1 | 247 | Intron | XP_016872245.1 | |||
XM_017016757.1 | 247 | UTR 5 | XP_016872246.1 | |||
XM_017016758.1 | 247 | UTR 5 | XP_016872247.1 |