Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615079 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASUN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASUN - asunder, spermatogenesis regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018164.2 | 1894 | Silent Mutation | GAA,GAG | E,E 608 | NP_060634.2 | |
XM_005253422.3 | 1894 | Silent Mutation | GAA,GAG | E,E 551 | XP_005253479.1 | |
XM_011520750.2 | 1894 | Silent Mutation | GAA,GAG | E,E 614 | XP_011519052.1 | |
XM_011520751.2 | 1894 | Silent Mutation | GAA,GAG | E,E 557 | XP_011519053.1 | |
XM_011520752.2 | 1894 | Silent Mutation | GAA,GAG | E,E 539 | XP_011519054.1 | |
XM_011520753.2 | 1894 | Silent Mutation | GAA,GAG | E,E 336 | XP_011519055.1 | |
XM_017019632.1 | 1894 | Silent Mutation | GAA,GAG | E,E 608 | XP_016875121.1 | |
XM_017019633.1 | 1894 | Silent Mutation | GAA,GAG | E,E 589 | XP_016875122.1 | |
XM_017019634.1 | 1894 | Silent Mutation | GAA,GAG | E,E 583 | XP_016875123.1 | |
XM_017019635.1 | 1894 | Silent Mutation | GAA,GAG | E,E 551 | XP_016875124.1 | |
XM_017019636.1 | 1894 | Silent Mutation | GAA,GAG | E,E 533 | XP_016875125.1 | |
XM_017019637.1 | 1894 | Silent Mutation | GAA,GAG | E,E 330 | XP_016875126.1 |