Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCGGGAACCCACTCAGAGCGAG[G/T]CTAAATTTACGGAGGGACTTTCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603021 MIM: 612804 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPS12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MRPS12 - mitochondrial ribosomal protein S12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021107.1 | 158 | UTR 5 | NP_066930.1 | |||
NM_033362.3 | 158 | Intron | NP_203526.1 | |||
NM_033363.1 | 158 | UTR 5 | NP_203527.1 |
SARS2 - seryl-tRNA synthetase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145901.1 | 158 | Intron | NP_001139373.1 | |||
NM_017827.3 | 158 | Intron | NP_060297.1 |
Set Membership: |
HapMap |