Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGGCACTGCCCAGGACCCCACCC[C/T]AGAAGCCCCTCCTCACCCTCGGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611262 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOHH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DOHH - deoxyhypusine hydroxylase/monooxygenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145165.1 | Intron | NP_001138637.1 | ||||
NM_031304.4 | Intron | NP_112594.1 | ||||
XM_011528341.2 | Intron | XP_011526643.1 |
Set Membership: |
Validated |