Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGACCACTCTGGCACCTTGAGGAG[A/G]AGCCAATCTGACAGGACCGAATACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF38 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARHGEF38 - Rho guanine nucleotide exchange factor 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242729.1 | 39 | Silent Mutation | AGA,AGG | R,R 52 | NP_001229658.1 | |
NM_017700.1 | 39 | Silent Mutation | AGA,AGG | R,R 52 | NP_060170.1 | |
XM_006714244.3 | 39 | Intron | XP_006714307.1 | |||
XM_011532050.2 | 39 | UTR 5 | XP_011530352.1 | |||
XM_011532051.2 | 39 | Silent Mutation | AGA,AGG | R,R 52 | XP_011530353.1 |
ARHGEF38-IT1 - ARHGEF38 intronic transcript 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |