Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164765 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTTN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CTTN - cortactin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184740.1 | 278 | Silent Mutation | ACC,ACT | T,T 24 | NP_001171669.1 | |
NM_005231.3 | 278 | Silent Mutation | ACC,ACT | T,T 24 | NP_005222.2 | |
NM_138565.2 | 278 | Silent Mutation | ACC,ACT | T,T 24 | NP_612632.1 | |
XM_006718447.2 | 278 | Silent Mutation | ACC,ACT | T,T 24 | XP_006718510.1 | |
XM_006718448.3 | 278 | Silent Mutation | ACC,ACT | T,T 24 | XP_006718511.1 | |
XM_017017312.1 | 278 | Silent Mutation | ACC,ACT | T,T 24 | XP_016872801.1 |
Set Membership: |
HapMap |