Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 159465 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MOG PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MOG - myelin oligodendrocyte glycoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008228.2 | Intron | NP_001008229.1 | ||||
NM_001008229.2 | Intron | NP_001008230.1 | ||||
NM_001170418.1 | Intron | NP_001163889.1 | ||||
NM_002433.4 | Intron | NP_002424.3 | ||||
NM_206809.3 | Intron | NP_996532.2 | ||||
NM_206810.3 | Intron | NP_996533.2 | ||||
NM_206811.3 | Intron | NP_996534.2 | ||||
NM_206812.3 | Intron | NP_996535.2 | ||||
NM_206814.5 | Intron | NP_996537.3 | ||||
XM_005249131.3 | Intron | XP_005249188.1 |
Set Membership: |
HapMap |