Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAAGGGAGAGAAAGATTACAAAC[C/T]GAGGTGTGGGCAGATGGAGACAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601393 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCL15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCL15 - C-C motif chemokine ligand 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032965.5 | Intron | NP_116741.2 |
CCL15-CCL14 - CCL15-CCL14 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101927339 - uncharacterized LOC101927339 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |