Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTAGAGCTGAAGGACGATCGCCC[G/T]GCCAGAGATATGTGGGTAATGGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107265 MIM: 611869 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD19 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CD19 - CD19 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178098.1 | 568 | Silent Mutation | CCG,CCT | P,P 235 | NP_001171569.1 | |
NM_001770.5 | 568 | Silent Mutation | CCG,CCT | P,P 235 | NP_001761.3 | |
XM_006721103.3 | 568 | Silent Mutation | CCG,CCT | P,P 146 | XP_006721166.1 | |
XM_011545981.1 | 568 | Silent Mutation | CCG,CCT | P,P 235 | XP_011544283.1 | |
XM_017023893.1 | 568 | Silent Mutation | CCG,CCT | P,P 146 | XP_016879382.1 |
RABEP2 - rabaptin, RAB GTPase binding effector protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |