Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116955 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNBP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNBP - CCHC-type zinc finger nucleic acid binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127192.1 | 362 | Silent Mutation | NP_001120664.1 | |||
NM_001127193.1 | 362 | Silent Mutation | NP_001120665.1 | |||
NM_001127194.1 | 362 | Silent Mutation | NP_001120666.1 | |||
NM_001127195.1 | 362 | Silent Mutation | NP_001120667.1 | |||
NM_001127196.1 | 362 | Silent Mutation | NP_001120668.1 | |||
NM_003418.4 | 362 | Silent Mutation | NP_003409.1 |