Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCCCCTTGGGATCCCTCCATCCC[G/T]CCATCCCTCGTGCTGCAGTATCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605418 MIM: 601729 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DKKL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DKKL1 - dickkopf like acrosomal protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197301.1 | Intron | NP_001184230.1 | ||||
NM_001197302.1 | Intron | NP_001184231.1 | ||||
NM_014419.3 | Intron | NP_055234.1 | ||||
XM_006723142.2 | Intron | XP_006723205.1 | ||||
XM_011526724.1 | Intron | XP_011525026.1 | ||||
XM_011526725.1 | Intron | XP_011525027.1 | ||||
XM_011526726.2 | Intron | XP_011525028.1 | ||||
XM_011526727.2 | Intron | XP_011525029.1 | ||||
XM_011526728.2 | Intron | XP_011525030.1 | ||||
XM_011526729.1 | Intron | XP_011525031.1 | ||||
XM_017026593.1 | Intron | XP_016882082.1 |
LOC101928295 - uncharacterized LOC101928295 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEAD2 - TEA domain transcription factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |