Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCAAGCGGACAGCCCCGCGCCGC[C/T]TCCACCTTTGCGGGTTTTCCCGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601955 MIM: 611996 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDK7 - cyclin dependent kinase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324069.1 | Intron | NP_001310998.1 | ||||
NM_001324070.1 | Intron | NP_001310999.1 | ||||
NM_001324071.1 | Intron | NP_001311000.1 | ||||
NM_001324072.1 | Intron | NP_001311001.1 | ||||
NM_001324074.1 | Intron | NP_001311003.1 | ||||
NM_001324075.1 | Intron | NP_001311004.1 | ||||
NM_001324077.1 | Intron | NP_001311006.1 | ||||
NM_001324078.1 | Intron | NP_001311007.1 | ||||
NM_001799.3 | Intron | NP_001790.1 | ||||
XM_011543094.2 | Intron | XP_011541396.1 |
MRPS36 - mitochondrial ribosomal protein S36 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |