Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 311550 MIM: 314370 MIM: 300050 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK16 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CDK16 - cyclin dependent kinase 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170460.1 | 1267 | Silent Mutation | TCA,TCG | S,S 193 | NP_001163931.1 | |
NM_006201.4 | 1267 | Silent Mutation | TCA,TCG | S,S 119 | NP_006192.1 | |
NM_033018.3 | 1267 | Silent Mutation | TCA,TCG | S,S 125 | NP_148978.2 | |
XM_011543920.2 | 1267 | Silent Mutation | TCA,TCG | S,S 193 | XP_011542222.1 | |
XM_011543921.1 | 1267 | Silent Mutation | TCA,TCG | S,S 166 | XP_011542223.1 | |
XM_011543922.1 | 1267 | Silent Mutation | TCA,TCG | S,S 162 | XP_011542224.1 | |
XM_011543923.1 | 1267 | Silent Mutation | TCA,TCG | S,S 125 | XP_011542225.1 | |
XM_011543924.1 | 1267 | Silent Mutation | TCA,TCG | S,S 119 | XP_011542226.1 | |
XM_011543925.2 | 1267 | Silent Mutation | TCA,TCG | S,S 119 | XP_011542227.1 | |
XM_011543926.1 | 1267 | Silent Mutation | TCA,TCG | S,S 119 | XP_011542228.1 | |
XM_011543927.2 | 1267 | Silent Mutation | TCA,TCG | S,S 119 | XP_011542229.1 | |
XM_011543928.1 | 1267 | Silent Mutation | TCA,TCG | S,S 119 | XP_011542230.1 | |
XM_017029569.1 | 1267 | Silent Mutation | TCA,TCG | S,S 166 | XP_016885058.1 | |
XM_017029570.1 | 1267 | Silent Mutation | TCA,TCG | S,S 119 | XP_016885059.1 | |
XM_017029571.1 | 1267 | Silent Mutation | TCA,TCG | S,S 119 | XP_016885060.1 | |
XM_017029572.1 | 1267 | Silent Mutation | TCA,TCG | S,S 119 | XP_016885061.1 | |
XM_017029573.1 | 1267 | Silent Mutation | TCA,TCG | S,S 119 | XP_016885062.1 |
UBA1 - ubiquitin like modifier activating enzyme 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP11 - ubiquitin specific peptidase 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |