Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGTTGAGAAGCACGCCCACCCG[C/T]GTGGCCTTGGTGGAGGGCAGGGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600453 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR3614 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR3614 - microRNA 3614 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTVR2 - mouse mammary tumor virus receptor homolog 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM25 - tripartite motif containing 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005082.4 | 1785 | Silent Mutation | ACA,ACG | T,T 575 | NP_005073.2 |