Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601601 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TFAP2B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TFAP2B - transcription factor AP-2 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003221.3 | 405 | UTR 5 | NP_003212.2 | |||
XM_011514837.2 | 405 | UTR 5 | XP_011513139.1 | |||
XM_017011233.1 | 405 | Missense Mutation | AAT,AGT | N,S 20 | XP_016866722.1 | |
XM_017011234.1 | 405 | Missense Mutation | AAT,AGT | N,S 8 | XP_016866723.1 | |
XM_017011235.1 | 405 | UTR 5 | XP_016866724.1 |
Set Membership: |
HapMap |