Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGAAGCGAGTCTGAGGCCTGGGG[C/T]TGCTTTTCTCCCTACACCTGTCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TP53I11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TP53I11 - tumor protein p53 inducible protein 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258320.1 | 2498 | UTR 3 | NP_001245249.1 | |||
NM_001258321.1 | 2498 | UTR 3 | NP_001245250.1 | |||
NM_001258322.1 | 2498 | UTR 3 | NP_001245251.1 | |||
NM_001258323.2 | 2498 | UTR 3 | NP_001245252.1 | |||
NM_001258324.1 | 2498 | UTR 3 | NP_001245253.1 | |||
NM_001318384.1 | 2498 | UTR 3 | NP_001305313.1 | |||
NM_001318385.1 | 2498 | UTR 3 | NP_001305314.1 | |||
NM_001318386.1 | 2498 | UTR 3 | NP_001305315.1 | |||
NM_001318387.1 | 2498 | UTR 3 | NP_001305316.1 | |||
NM_001318388.1 | 2498 | UTR 3 | NP_001305317.1 | |||
NM_001318389.1 | 2498 | UTR 3 | NP_001305318.1 | |||
NM_001318390.1 | 2498 | UTR 3 | NP_001305319.1 | |||
NM_006034.4 | 2498 | UTR 3 | NP_006025.2 | |||
XM_005253227.4 | 2498 | UTR 3 | XP_005253284.1 | |||
XM_005253229.1 | 2498 | Intron | XP_005253286.1 | |||
XM_011520475.2 | 2498 | UTR 3 | XP_011518777.1 | |||
XM_011520476.2 | 2498 | UTR 3 | XP_011518778.1 | |||
XM_011520477.2 | 2498 | UTR 3 | XP_011518779.1 | |||
XM_011520478.2 | 2498 | UTR 3 | XP_011518780.1 | |||
XM_017018580.1 | 2498 | UTR 3 | XP_016874069.1 | |||
XM_017018581.1 | 2498 | UTR 3 | XP_016874070.1 |
TSPAN18 - tetraspanin 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130783.4 | 2498 | Intron | NP_570139.3 | |||
XM_005253217.3 | 2498 | Intron | XP_005253274.1 | |||
XM_006718372.3 | 2498 | Intron | XP_006718435.1 | |||
XM_006718373.3 | 2498 | Intron | XP_006718436.1 | |||
XM_011520459.2 | 2498 | Intron | XP_011518761.1 | |||
XM_017018530.1 | 2498 | Intron | XP_016874019.1 | |||
XM_017018531.1 | 2498 | Intron | XP_016874020.1 |