Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCACCCAGTTCCCCATTACAGC[A/C]TTGCGGGAGATCAAGATCCTTCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603251 MIM: 613405 MIM: 604722 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDK9 - cyclin dependent kinase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261.3 | 312 | Silent Mutation | GCA,GCC | A,A 63 | NP_001252.1 | |
XM_017014184.1 | 312 | UTR 5 | XP_016869673.1 |
MIR2861 - microRNA 2861 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3960 - microRNA 3960 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH2D3C - SH2 domain containing 3C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |