Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTCTCAGCAAGCTTCAGGGCTTC[A/G]TGCATGCCTGCAGCTGAGAGCTTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 182137 | ||||||||||||||||||||
Literature Links: |
HTR7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HTR7 - 5-hydroxytryptamine receptor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000872.4 | 1491 | Silent Mutation | CAC,CAT | H,H 416 | NP_000863.1 | |
NM_019859.3 | 1491 | Silent Mutation | CAC,CAT | H,H 416 | NP_062873.1 | |
NM_019860.3 | 1491 | Silent Mutation | CAC,CAT | H,H 416 | NP_062874.1 |