Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612982 MIM: 143023 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LMNTD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LMNTD2 - lamin tail domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC56 - leucine rich repeat containing 56 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR210 - microRNA 210 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR210HG - MIR210 host gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASSF7 - Ras association domain family member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143993.1 | Intron | NP_001137465.1 | ||||
NM_001143994.1 | Intron | NP_001137466.1 | ||||
NM_003475.3 | Intron | NP_003466.1 | ||||
XM_005253149.3 | Intron | XP_005253206.2 | ||||
XM_005253150.1 | Intron | XP_005253207.1 | ||||
XM_006718338.2 | Intron | XP_006718401.1 | ||||
XM_011520389.1 | Intron | XP_011518691.1 | ||||
XM_017018360.1 | Intron | XP_016873849.1 | ||||
XM_017018361.1 | Intron | XP_016873850.1 | ||||
XM_017018362.1 | Intron | XP_016873851.1 |