Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCACCTCTGGGCAGAGAAACAAAG[C/T]TCTATATGCACAGCCCAGCAAAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606534 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP3A43 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CYP3A43 - cytochrome P450 family 3 subfamily A member 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278921.1 | 23 | UTR 5 | NP_001265850.1 | |||
NM_022820.4 | 23 | UTR 5 | NP_073731.1 | |||
NM_057095.2 | 23 | UTR 5 | NP_476436.1 | |||
NM_057096.3 | 23 | UTR 5 | NP_476437.1 | |||
XM_017012542.1 | 23 | Intron | XP_016868031.1 | |||
XM_017012543.1 | 23 | Intron | XP_016868032.1 | |||
XM_017012544.1 | 23 | Intron | XP_016868033.1 | |||
XM_017012545.1 | 23 | Intron | XP_016868034.1 |
Set Membership: |
DME Validated Inventoried |