Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCATTGAATGAATTTAAATCATC[A/T]TCTTTGGCCTGAAATAAATGTTACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151570 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LTA4H PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LTA4H - leukotriene A4 hydrolase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000895.2 | 1575 | Missense Mutation | GAA,GAT | E,D 481 | NP_000886.1 | |
NM_001256643.1 | 1575 | Missense Mutation | GAA,GAT | E,D 457 | NP_001243572.1 | |
NM_001256644.1 | 1575 | Missense Mutation | GAA,GAT | E,D 457 | NP_001243573.1 | |
XM_005268871.1 | 1575 | Missense Mutation | GAA,GAT | E,D 481 | XP_005268928.1 | |
XM_011538348.1 | 1575 | Missense Mutation | GAA,GAT | E,D 481 | XP_011536650.1 | |
XM_011538349.2 | 1575 | Intron | XP_011536651.1 |