Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTCCCAGCTAGTCCACAGGGACC[A/G]GCAGTGCCATGGAAGGATTTAACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609601 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF397 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF397 - zinc finger protein 397 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135178.2 | 594 | Silent Mutation | CCA,CCG | P,P 146 | NP_001128650.1 | |
NM_032347.2 | 594 | Silent Mutation | CCA,CCG | P,P 146 | NP_115723.1 | |
XM_006722558.2 | 594 | Silent Mutation | CCA,CCG | P,P 165 | XP_006722621.1 | |
XM_011526229.2 | 594 | Silent Mutation | CCA,CCG | P,P 165 | XP_011524531.1 | |
XM_011526230.2 | 594 | Silent Mutation | CCA,CCG | P,P 165 | XP_011524532.1 | |
XM_011526231.2 | 594 | Silent Mutation | CCA,CCG | P,P 146 | XP_011524533.1 | |
XM_017026042.1 | 594 | Silent Mutation | CCA,CCG | P,P 146 | XP_016881531.1 |
ZSCAN30 - zinc finger and SCAN domain containing 30 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |