Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602757 MIM: 606680 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPHB6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EPHB6 - EPH receptor B6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001280794.2 | 2336 | Intron | NP_001267723.2 | |||
NM_001280795.2 | 2336 | Intron | NP_001267724.2 | |||
NM_004445.5 | 2336 | Intron | NP_004436.4 | |||
XM_006715881.2 | 2336 | Intron | XP_006715944.1 | |||
XM_011515879.1 | 2336 | Intron | XP_011514181.1 | |||
XM_011515880.1 | 2336 | Intron | XP_011514182.1 | |||
XM_011515881.2 | 2336 | Intron | XP_011514183.1 | |||
XM_011515882.2 | 2336 | Intron | XP_011514184.1 |
TRPV6 - transient receptor potential cation channel subfamily V member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018646.5 | 2336 | Silent Mutation | CTG,TTG | L,L 743 | NP_061116.5 |
Set Membership: |
HapMap |