Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTACGAGGAATACTCTTTTCACTG[C/T]CAAATAACTCTCAGCTCATCTGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611604 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERLIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERLIN1 - ER lipid raft associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100626.1 | Intron | NP_001094096.1 | ||||
NM_006459.3 | Intron | NP_006450.2 | ||||
XM_005269442.3 | Intron | XP_005269499.1 | ||||
XM_017015541.1 | Intron | XP_016871030.1 |