Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGGCTAGCGCCCCTGCCCCCTC[A/G]GCCAGCCCGCCTGCACTGCCACCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611888 MIM: 606784 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERF PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ERF - ETS2 repressor factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301035.1 | 1255 | Silent Mutation | GCC,GCT | A,A 335 | NP_001287964.1 | |
NM_001308402.1 | 1255 | Silent Mutation | GCC,GCT | A,A 335 | NP_001295331.1 | |
NM_001312656.1 | 1255 | Silent Mutation | GCC,GCT | A,A 335 | NP_001299585.1 | |
NM_006494.3 | 1255 | Silent Mutation | GCC,GCT | A,A 410 | NP_006485.2 | |
XM_017026468.1 | 1255 | Silent Mutation | GCC,GCT | A,A 335 | XP_016881957.1 | |
XM_017026469.1 | 1255 | Silent Mutation | GCC,GCT | A,A 335 | XP_016881958.1 |
GSK3A - glycogen synthase kinase 3 alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |