Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGAAGTATAATAGCAGTTCTTTT[A/C]TCTTTATAGGTTAGACCAAAGCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164785 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100130075 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100130075 - SUZ RNA binding domain containing 1 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MDM2 - MDM2 proto-oncogene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145337.2 | Intron | NP_001138809.1 | ||||
NM_001145339.2 | Intron | NP_001138811.1 | ||||
NM_001145340.2 | Intron | NP_001138812.1 | ||||
NM_001278462.1 | Intron | NP_001265391.1 | ||||
NM_002392.5 | Intron | NP_002383.2 | ||||
XM_005268872.4 | Intron | XP_005268929.1 | ||||
XM_006719399.3 | Intron | XP_006719462.1 | ||||
XM_006719400.3 | Intron | XP_006719463.1 |