Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCAGGACTGACACCACAGTTTTG[G/A]GGTAAGCAGCTGTGGAAGGGTGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300272 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDAC6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HDAC6 - histone deacetylase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321225.1 | Intron | NP_001308154.1 | ||||
NM_001321226.1 | Intron | NP_001308155.1 | ||||
NM_001321227.1 | Intron | NP_001308156.1 | ||||
NM_001321228.1 | Intron | NP_001308157.1 | ||||
NM_001321229.1 | Intron | NP_001308158.1 | ||||
NM_001321230.1 | Intron | NP_001308159.1 | ||||
NM_001321231.1 | Intron | NP_001308160.1 | ||||
NM_006044.3 | Intron | NP_006035.2 | ||||
XM_017029187.1 | Intron | XP_016884676.1 |