Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGGCCTGCCCGCTCCTGTCTATC[C/T]GCAGGTCTTCCTTCAGGCCTGGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601978 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARID3C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARID3C - AT-rich interaction domain 3C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIGMAR1 - sigma non-opioid intracellular receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282205.1 | 851 | UTR 3 | NP_001269134.1 | |||
NM_001282206.1 | 851 | UTR 3 | NP_001269135.1 | |||
NM_001282207.1 | 851 | UTR 3 | NP_001269136.1 | |||
NM_001282208.1 | 851 | UTR 3 | NP_001269137.1 | |||
NM_001282209.1 | 851 | UTR 3 | NP_001269138.1 | |||
NM_005866.3 | 851 | UTR 3 | NP_005857.1 | |||
NM_147157.2 | 851 | UTR 3 | NP_671513.1 |