Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCACTTCAGGCTTGTGATGTCCCT[G/A]TTGAGGAGGGGCCAGTCCCCAGAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
GLOD5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLOD5 - glyoxalase domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080489.2 | 517 | Missense Mutation | ATT,GTT | I,V 125 | NP_001073958.2 | |
XM_011543911.2 | 517 | Missense Mutation | ATT,GTT | I,V 83 | XP_011542213.1 | |
XM_011543912.2 | 517 | Missense Mutation | ATT,GTT | I,V 148 | XP_011542214.1 |