Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCAAGCTGGACGTGGGAGAGTG[A/G]AGCACTACCTCCTCAGGCCAGGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611981 MIM: 612648 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPS18A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPS18A - mitochondrial ribosomal protein S18A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193343.1 | Intron | NP_001180272.1 | ||||
NM_018135.3 | Intron | NP_060605.1 | ||||
XM_006715134.2 | Intron | XP_006715197.1 |
RSPH9 - radial spoke head 9 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193341.1 | Intron | NP_001180270.1 | ||||
NM_152732.4 | Intron | NP_689945.2 | ||||
XM_005248901.3 | Intron | XP_005248958.1 | ||||
XM_006715014.1 | Intron | XP_006715077.1 | ||||
XM_017010426.1 | Intron | XP_016865915.1 |