Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGATCATCCACGACGACCACCC[T/G]CTGTCCTGGCTCCAGGGCGTCTTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 102600 MIM: 605525 MIM: 612222 | ||||||||||||||||||||
Literature Links: |
APRT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APRT - adenine phosphoribosyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000485.2 | 399 | Silent Mutation | AGG,CGG | R,R 122 | NP_000476.1 | |
NM_001030018.1 | 399 | Silent Mutation | AGG,CGG | R,R 122 | NP_001025189.1 |
CDT1 - chromatin licensing and DNA replication factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030928.3 | 399 | Intron | NP_112190.2 |
GALNS - galactosamine (N-acetyl)-6-sulfatase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |