Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTAGAAAGGAGTCCTAGGGGAT[A/G]ATTCTGTTTTCCCATGAGTTATGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602518 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LGALS4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LGALS4 - galectin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006149.3 | 1629 | UTR 3 | NP_006140.1 | |||
XM_011526973.1 | 1629 | UTR 3 | XP_011525275.1 | |||
XM_011526974.1 | 1629 | UTR 3 | XP_011525276.1 |
LGALS7B - galectin 7B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |