Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGGCCACCCCCCAGCGCTGTTGC[G/A]TTGGCTAGAGGGCGCTTTTCCATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600693 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4745 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR4745 - microRNA 4745 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTBP1 - polypyrimidine tract binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002819.4 | Intron | NP_002810.1 | ||||
NM_031990.3 | Intron | NP_114367.1 | ||||
NM_031991.3 | Intron | NP_114368.1 | ||||
XM_005259597.2 | Intron | XP_005259654.1 | ||||
XM_005259598.2 | Intron | XP_005259655.1 |