Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCACTGAAGGGGGAGATGGTTACT[G/T]TCCGGCCGACGGGCAGGATGGGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300102 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PNPLA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PNPLA4 - patatin like phospholipase domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142389.1 | 600 | Missense Mutation | AAA,ACA | K,T 178 | NP_001135861.1 | |
NM_001172672.1 | 600 | Missense Mutation | AAA,ACA | K,T 91 | NP_001166143.1 | |
NM_004650.2 | 600 | Missense Mutation | AAA,ACA | K,T 178 | NP_004641.1 | |
XM_011545588.1 | 600 | Missense Mutation | AAA,ACA | K,T 178 | XP_011543890.1 |