Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGCCTGCCCCAGCTGCTCACCT[C/T]CCCTTTCCCACAGAGCAGGGCCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604077 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF135 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF135 - zinc finger protein 135 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164527.1 | 415 | Intron | NP_001157999.1 | |||
NM_001164529.1 | 415 | Intron | NP_001158001.1 | |||
NM_001164530.1 | 415 | Intron | NP_001158002.1 | |||
NM_001289401.1 | 415 | Intron | NP_001276330.1 | |||
NM_001289402.1 | 415 | Intron | NP_001276331.1 | |||
NM_003436.3 | 415 | Intron | NP_003427.3 | |||
NM_007134.1 | 415 | Intron | NP_009065.1 | |||
XM_005259211.1 | 415 | Intron | XP_005259268.1 | |||
XM_006723362.3 | 415 | Intron | XP_006723425.1 | |||
XM_006723363.3 | 415 | Intron | XP_006723426.1 | |||
XM_017027240.1 | 415 | UTR 5 | XP_016882729.1 | |||
XM_017027241.1 | 415 | Intron | XP_016882730.1 | |||
XM_017027242.1 | 415 | Intron | XP_016882731.1 |
ZSCAN1 - zinc finger and SCAN domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |