Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGCTTAGAAGTCTGTACTACTGC[A/T]CCCTGCAAAAAGCCTCAGAACTAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608342 MIM: 603142 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYB5R2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CYB5R2 - cytochrome b5 reductase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302826.1 | Intron | NP_001289755.1 | ||||
NM_001302827.1 | Intron | NP_001289756.1 | ||||
NM_016229.4 | Intron | NP_057313.2 | ||||
XM_005252975.4 | Intron | XP_005253032.1 | ||||
XM_006718251.2 | Intron | XP_006718314.1 | ||||
XM_011520184.2 | Intron | XP_011518486.1 | ||||
XM_011520185.2 | Intron | XP_011518487.1 | ||||
XM_017017920.1 | Intron | XP_016873409.1 | ||||
XM_017017921.1 | Intron | XP_016873410.1 | ||||
XM_017017922.1 | Intron | XP_016873411.1 |
PPFIBP2 - PPFIA binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256568.1 | Intron | NP_001243497.1 | ||||
NM_001256569.1 | Intron | NP_001243498.1 | ||||
NM_003621.3 | Intron | NP_003612.2 | ||||
XM_005253177.2 | Intron | XP_005253234.1 | ||||
XM_006718349.2 | Intron | XP_006718412.1 | ||||
XM_006718350.2 | Intron | XP_006718413.1 | ||||
XM_006718351.2 | Intron | XP_006718414.1 | ||||
XM_006718352.2 | Intron | XP_006718415.1 | ||||
XM_006718353.2 | Intron | XP_006718416.1 | ||||
XM_011520413.2 | Intron | XP_011518715.1 | ||||
XM_011520414.2 | Intron | XP_011518716.1 | ||||
XM_011520415.2 | Intron | XP_011518717.1 | ||||
XM_011520416.2 | Intron | XP_011518718.1 | ||||
XM_011520417.2 | Intron | XP_011518719.1 | ||||
XM_011520418.2 | Intron | XP_011518720.1 | ||||
XM_011520419.2 | Intron | XP_011518721.1 | ||||
XM_011520420.2 | Intron | XP_011518722.1 | ||||
XM_017018443.1 | Intron | XP_016873932.1 | ||||
XM_017018444.1 | Intron | XP_016873933.1 | ||||
XM_017018445.1 | Intron | XP_016873934.1 | ||||
XM_017018446.1 | Intron | XP_016873935.1 | ||||
XM_017018447.1 | Intron | XP_016873936.1 |
Set Membership: |
HapMap |