Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCCACGGACACTTCAGGCTATGG[C/T]CAGAGCAGCTATTCTTCTTATGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137070 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FUS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FUS - FUS RNA binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170634.1 | 924 | Silent Mutation | GGC,GGT | G,G 51 | NP_001164105.1 | |
NM_001170937.1 | 924 | Silent Mutation | GGC,GGT | G,G 51 | NP_001164408.1 | |
NM_004960.3 | 924 | Silent Mutation | GGC,GGT | G,G 51 | NP_004951.1 | |
XM_005255233.4 | 924 | UTR 5 | XP_005255290.1 | |||
XM_011545781.1 | 924 | Silent Mutation | GGC,GGT | G,G 51 | XP_011544083.1 | |
XM_011545782.2 | 924 | Intron | XP_011544084.1 |