Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCTCAGGGACCCCTTCTGGGGG[T/C]CCGGCAGGTGACACAGGGCGGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605847 MIM: 600435 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL7C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCL7C - BCL tumor suppressor 7C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286526.1 | 986 | Silent Mutation | GGA,GGG | G,G 129 | NP_001273455.1 | |
NM_004765.3 | 986 | Silent Mutation | GGA,GGG | G,G 129 | NP_004756.2 | |
XM_011545980.2 | 986 | Silent Mutation | GGA,GGG | G,G 129 | XP_011544282.1 | |
XM_017023885.1 | 986 | Silent Mutation | GGA,GGG | G,G 129 | XP_016879374.1 | |
XM_017023886.1 | 986 | Silent Mutation | GGA,GGG | G,G 129 | XP_016879375.1 | |
XM_017023887.1 | 986 | Silent Mutation | GGA,GGG | G,G 129 | XP_016879376.1 | |
XM_017023888.1 | 986 | Silent Mutation | GGA,GGG | G,G 129 | XP_016879377.1 |
CTF1 - cardiotrophin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR762 - microRNA 762 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR762HG - MIR762 host gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |