Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGGGTGACTGAGTGCCTCCGAG[G/T]GAAGCCCCCACCAGCAGGGGTGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601064 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFP36L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZFP36L1 - ZFP36 ring finger protein like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244698.1 | 1124 | Missense Mutation | ACT,CCT | T,P 50 | NP_001231627.1 | |
NM_001244701.1 | 1124 | Missense Mutation | ACT,CCT | T,P 119 | NP_001231630.1 | |
NM_004926.3 | 1124 | Missense Mutation | ACT,CCT | T,P 50 | NP_004917.2 |