Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAATGATGGGACTGGATGCATGCT[A/G]TTATTTCATTTTCACTATTACCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614903 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNX16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNX16 - sorting nexin 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022133.3 | Intron | NP_071416.2 | ||||
NM_152836.2 | Intron | NP_690049.1 | ||||
NM_152837.2 | Intron | NP_690050.1 | ||||
XM_005251282.4 | Intron | XP_005251339.1 | ||||
XM_005251283.2 | Intron | XP_005251340.1 | ||||
XM_011517574.2 | Intron | XP_011515876.1 |