Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCGGCCCTTCAGGCTCTGTCTCT[A/G]TGGAGACTGGGCTTTGGGAGGGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609998 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HINT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HINT3 - histidine triad nucleotide binding protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRMT11 - tRNA methyltransferase 11 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031712.2 | Intron | NP_001026882.2 | ||||
XM_017011168.1 | Intron | XP_016866657.1 | ||||
XM_017011169.1 | Intron | XP_016866658.1 |