Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGCAGGTAGTAATGGGTCATGTC[A/G]TAGAGGACGTAGCCCAGGAGGCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611026 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FA2H PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FA2H - fatty acid 2-hydroxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024306.4 | 708 | Silent Mutation | TAC,TAT | Y,Y 311 | NP_077282.3 | |
XM_011523317.2 | 708 | UTR 3 | XP_011521619.1 | |||
XM_011523319.2 | 708 | Silent Mutation | TAC,TAT | Y,Y 231 | XP_011521621.1 |
Set Membership: |
HapMap |