Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTGGGACCAGCGCCATCACCTCC[A/G]TACGGATGTTCTCCCTCCGGAAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601240 MIM: 601825 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAMT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GAMT - guanidinoacetate N-methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000156.5 | 720 | Missense Mutation | ACG,ATG | T,M 209 | NP_000147.1 | |
NM_138924.2 | 720 | Intron | NP_620279.1 |
NDUFS7 - NADH:ubiquinone oxidoreductase core subunit S7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |