Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGGTGGGGGCCTACATGGAGGAG[A/G]TGGGCTAGGGTCCCGGGGTGGTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612034 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APC2 - APC2, WNT signaling pathway regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005883.2 | Intron | NP_005874.1 | ||||
XM_005259475.2 | Intron | XP_005259532.1 | ||||
XM_006722607.2 | Intron | XP_006722670.1 | ||||
XM_006722608.3 | Intron | XP_006722671.2 | ||||
XM_006722609.3 | Intron | XP_006722672.1 | ||||
XM_006722610.3 | Intron | XP_006722673.2 |