Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTATAGGTATTATATTTTTGTCC[C/G]GGTTTTAAATCTGGAGTAAAGCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
28 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603478 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
USP1 - ubiquitin specific peptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017415.1 | 637 | Silent Mutation | CCC,CCG | P,P 103 | NP_001017415.1 | |
NM_001017416.1 | 637 | Silent Mutation | CCC,CCG | P,P 103 | NP_001017416.1 | |
NM_003368.4 | 637 | Silent Mutation | CCC,CCG | P,P 103 | NP_003359.3 |