Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGCTGAGAGGTGAGACCCCCAT[C/G]CCATCCACTGAACAGGAGACCCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 136425 MIM: 600829 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOLR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FOLR2 - folate receptor beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
INPPL1 - inositol polyphosphate phosphatase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001567.3 | Intron | NP_001558.3 | ||||
XM_005273978.3 | Intron | XP_005274035.1 | ||||
XM_005273979.4 | Intron | XP_005274036.1 | ||||
XM_011544999.2 | Intron | XP_011543301.1 | ||||
XM_011545000.2 | Intron | XP_011543302.1 | ||||
XM_017017673.1 | Intron | XP_016873162.1 |