Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCAAGGACTAAGAGGGGAGACAC[C/T]GCATGTGGAATATTCTGGCTGTGCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 136430 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOLR1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FOLR1 - folate receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000802.3 | Intron | NP_000793.1 | ||||
NM_016724.2 | Intron | NP_057936.1 | ||||
NM_016725.2 | Intron | NP_057937.1 | ||||
NM_016729.2 | Intron | NP_057941.1 |