Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614717 MIM: 613510 MIM: 612414 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANAPC15 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANAPC15 - anaphase promoting complex subunit 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278485.1 | Intron | NP_001265414.1 | ||||
NM_001278486.1 | Intron | NP_001265415.1 | ||||
NM_001278487.1 | Intron | NP_001265416.1 | ||||
NM_001278488.1 | Intron | NP_001265417.1 | ||||
NM_001278489.1 | Intron | NP_001265418.1 | ||||
NM_001278490.1 | Intron | NP_001265419.1 | ||||
NM_001278491.1 | Intron | NP_001265420.1 | ||||
NM_001278492.1 | Intron | NP_001265421.1 | ||||
NM_001278493.1 | Intron | NP_001265422.1 | ||||
NM_001278494.1 | Intron | NP_001265423.1 | ||||
NM_014042.2 | Intron | NP_054761.1 | ||||
XM_017017488.1 | Intron | XP_016872977.1 | ||||
XM_017017489.1 | Intron | XP_016872978.1 | ||||
XM_017017490.1 | Intron | XP_016872979.1 | ||||
XM_017017491.1 | Intron | XP_016872980.1 | ||||
XM_017017492.1 | Intron | XP_016872981.1 | ||||
XM_017017493.1 | Intron | XP_016872982.1 | ||||
XM_017017494.1 | Intron | XP_016872983.1 | ||||
XM_017017495.1 | Intron | XP_016872984.1 | ||||
XM_017017496.1 | Intron | XP_016872985.1 | ||||
XM_017017497.1 | Intron | XP_016872986.1 |
LAMTOR1 - late endosomal/lysosomal adaptor, MAPK and MTOR activator 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRTOMT - leucine rich transmembrane and O-methyltransferase domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145307.4 | Intron | NP_001138779.1 | ||||
NM_001145308.4 | Intron | NP_001138780.1 | ||||
NM_001145309.3 | Intron | NP_001138781.1 | ||||
NM_001145310.3 | Intron | NP_001138782.1 | ||||
NM_001205138.3 | Intron | NP_001192067.1 | ||||
NM_001271471.2 | Intron | NP_001258400.1 | ||||
NM_001318803.1 | Intron | NP_001305732.1 | ||||
NM_145309.5 | Intron | NP_660352.1 | ||||
XM_006718473.3 | Intron | XP_006718536.1 | ||||
XM_006718474.3 | Intron | XP_006718537.1 | ||||
XM_011544847.2 | Intron | XP_011543149.1 | ||||
XM_011544848.2 | Intron | XP_011543150.1 | ||||
XM_017017356.1 | Intron | XP_016872845.1 | ||||
XM_017017357.1 | Intron | XP_016872846.1 | ||||
XM_017017358.1 | Intron | XP_016872847.1 | ||||
XM_017017359.1 | Intron | XP_016872848.1 | ||||
XM_017017360.1 | Intron | XP_016872849.1 |
Set Membership: |
HapMap |