Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTTCTTCTGAGACACATCCAGC[A/C]CCCCCACGTCTCCCTCATATACACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605906 MIM: 606665 | ||||||||||||||||||||
Literature Links: |
LDB3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OPN4 - opsin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001030015.2 | 2536 | UTR 3 | NP_001025186.1 | |||
NM_033282.3 | 2536 | UTR 3 | NP_150598.1 | |||
XM_017016955.1 | 2536 | Intron | XP_016872444.1 | |||
XM_017016956.1 | 2536 | Intron | XP_016872445.1 | |||
XM_017016957.1 | 2536 | UTR 3 | XP_016872446.1 |